Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111277.1 | NP_001104747.1 | 4 | 401 | ||
NM_175135.4 | NP_780344.2 | 4 | 401 | |||
GenBank mRNA | AK145856.1 | - | 4 | 378 | ||
AK172173.1 | - | 4 | 395 | |||
AK172302.1 | - | 4 | 351 | |||
BC115657.1 | - | 2 | 75 | 40103205 | ||
BC115658.1 | - | 2 | 75 | 40103206 |