Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160353.1 | NP_001153825.1 | 3 | 1345 | ||
GenBank mRNA | BC115866.1 | - | 3 | 1318 | 40081345 |
Set Membership: |
G-protein coupled receptor |