Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grm6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173372.2 | NP_775548.2 | 9 | 2186 | ||
GenBank mRNA | BC034118.1 | - | 5 | 1187 | ||
BC041684.1 | - | 4 | 1195 | 4502870 |
Set Membership: |
![]() |