Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301094.1 | NP_001288023.1 | 2 | 212 | ||
NM_138648.2 | NP_619589.2 | 2 | 212 | |||
GenBank mRNA | AF303744.1 | - | 2 | 201 | ||
AK154687.1 | - | 2 | 213 | |||
AK160919.1 | - | 2 | 212 | |||
HQ603086.1 | - | 2 | 154 |