Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkaa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178143.2 | NP_835279.2 | 7 | 918 | ||
GenBank mRNA | AK044030.1 | - | 7 | 754 | ||
AK134573.1 | - | 7 | 776 | |||
BC138565.1 | - | 7 | 927 | 8861585 | ||
BC138566.1 | - | 7 | 927 | 8861586 | ||
FJ588504.1 | - | 6 | 676 |