Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038643.1 | NP_001033732.1 | 3 | 911 | ||
NM_023908.2 | NP_076397.2 | 3 | 911 | |||
GenBank mRNA | AF226324.1 | - | 3 | 809 | ||
AK031280.1 | - | 3 | 911 | |||
AK037938.1 | - | 3 | 726 | |||
AK046945.1 | - | 2 | 271 | |||
AK151781.1 | - | 3 | 881 | |||
AK152488.1 | - | 3 | 745 | |||
AK166266.1 | - | 3 | 682 | |||
BC025059.1 | - | 3 | 744 | 2598913 | ||
BC094464.1 | - | 3 | 703 |