Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema3d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028882.4 | NP_083158.3 | 18 | 2676 | ||
GenBank mRNA | AK028900.1 | - | 18 | 2458 | ||
AK031704.1 | - | 18 | 2288 | |||
AK052671.1 | - | 18 | 2677 | |||
AK163828.1 | - | 3 | 468 | |||
BC138131.1 | - | 18 | 2219 | 8861150 |