Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ogt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290535.1 | NP_001277464.1 | 17 | 2230 | ||
NM_139144.4 | NP_631883.2 | 17 | 2427 | |||
GenBank mRNA | AF363030.1 | - | 17 | 2192 | ||
AK045910.1 | - | 2 | 176 | |||
BC038435.1 | - | 13 | 2486 | |||
BC048239.1 | - | 13 | 2182 | |||
BC057319.1 | - | 17 | 2350 | 6830845 |