Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ogt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290535.1 | NP_001277464.1 | 11, 12 | 1468 | ||
NM_139144.4 | NP_631883.2 | 11, 12 | 1665 | |||
GenBank mRNA | AF363030.1 | - | 11, 12 | 1430 | ||
BC038435.1 | - | 7, 8 | 1724 | |||
BC048239.1 | - | 7, 8 | 1421 | |||
BC057319.1 | - | 11, 12 | 1588 | 6830845 |