Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mthfd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138745.2 | NP_620084.2 | 24, 25 | 2737 | ||
GenBank mRNA | AF364579.1 | - | 24, 25 | 2518 | ||
AK088700.1 | - | 24, 25 | 2534 | |||
AK129012.1 | - | 24, 25 | 2671 | |||
AK133848.1 | - | 23, 24 | 2472 | |||
AK159812.1 | - | 24, 25 | 2549 | |||
AK165951.1 | - | 24, 25 | 2572 | |||
AK186267.1 | - | 1, 2 | 154 | |||
BC008523.1 | - | 24, 25 | 2561 | 3590449 | ||
BC013563.1 | - | 18, 19 | 1874 |