Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epsti1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029495.2 | NP_083771.1 | 5 | 664 | ||
NM_178825.4 | NP_849147.1 | 5 | 664 | |||
GenBank mRNA | AK017174.1 | - | 5 | 497 | ||
AK077223.1 | - | 5 | 470 | |||
AK077376.1 | - | 5 | 499 | |||
AK135710.1 | - | 5 | 658 | |||
AK136055.1 | - | 5 | 665 | |||
AY239223.1 | - | 5 | 497 | |||
AY239224.1 | - | 5 | 570 | |||
BC020120.1 | - | 5 | 497 | 3988503 | ||
BC021821.1 | - | 5 | 524 | 3581940 |