Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023731.3 | NP_076220.2 | 1 | 153 | ||
GenBank mRNA | AB041663.1 | - | 1 | 122 | ||
AK036812.1 | - | 1 | 143 | |||
AK078407.1 | - | 1 | 154 | |||
AK078455.1 | - | 1 | 154 | |||
AK088727.1 | - | 1 | 146 | |||
AK162676.1 | - | 1 | 139 | |||
BC043326.1 | - | 1 | 149 | 5374087 | ||
DQ501252.1 | - | 1 | 120 |