Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173866.3 | NP_776291.1 | 6 | 790 | ||
GenBank mRNA | AK031958.1 | - | 6 | 982 | ||
AK033424.1 | - | 6 | 791 | |||
AK075894.1 | - | 6 | 809 | |||
AK076250.1 | - | 6 | 808 | |||
AK082030.1 | - | 6 | 833 | |||
AK206462.1 | - | 1 | 16 | |||
BC034219.1 | - | 6 | 822 | 5065322 |