Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Obfc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175360.2 | NP_780569.1 | 5 | 641 | ||
GenBank mRNA | AK075574.1 | - | 5 | 641 | ||
AK135228.1 | - | 5 | 659 | |||
AK146929.1 | - | 5 | 719 | |||
AK164386.1 | - | 5 | 651 | |||
BC029548.1 | - | 5 | 681 | 5012064 |