Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDN11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185056.1 | NP_001171985.1 | 3 | 1260 | ||
NM_005602.5 | NP_005593.2 | 3 | 1590 | |||
GenBank mRNA | AJ245902.1 | - | 1 | 124 | ||
BC013577.1 | - | 3 | 1566 | 3895040 |