Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oxsr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133985.2 | NP_598746.2 | 17 | 1835 | ||
GenBank mRNA | AK031790.1 | - | 16 | 1332 | ||
AK075837.1 | - | 16 | 1343 | |||
AK129291.1 | - | 17 | 1830 | |||
BC036148.1 | - | 8 | 585 | |||
BC046453.1 | - | 18 | 1859 | |||
BC060645.1 | - | 17 | 1818 | 6852021 |