Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ston2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175367.6 | NP_780576.1 | 5 | 2506 | ||
GenBank mRNA | AK036612.1 | - | 5 | 2507 | ||
BC132531.1 | - | 4 | 2092 | 40130808 | ||
BC138054.1 | - | 4 | 2092 | 8861073 |