Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atad3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_179203.3 | NP_849534.2 | 3 | 440 | ||
GenBank mRNA | AF343079.1 | - | 3 | 486 | ||
AK075921.1 | - | 3 | 436 | |||
AK148974.1 | - | 3 | 439 | |||
AK149702.1 | - | 3 | 439 | |||
AK150037.1 | - | 3 | 439 | |||
AK165724.1 | - | 3 | 437 | |||
AK166099.1 | - | 3 | 436 | |||
AK172038.1 | - | 3 | 436 | |||
AK172350.1 | - | 3 | 441 | |||
AK173141.1 | - | 3 | 388 | |||
AK185628.1 | - | 3 | 168 | |||
BC023301.1 | - | 3 | 377 | 3671787 | ||
BC047047.1 | - | 3 | 403 | |||
BC058373.1 | - | 3 | 424 | 6828037 | ||
BC060036.1 | - | 3 | 372 | 6530155 |