Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atad3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_179203.3 | NP_849534.2 | 8 | 858 | ||
GenBank mRNA | AF343079.1 | - | 8 | 904 | ||
AK075921.1 | - | 8 | 854 | |||
AK148974.1 | - | 8 | 857 | |||
AK149702.1 | - | 8 | 857 | |||
AK150037.1 | - | 8 | 857 | |||
AK165724.1 | - | 8 | 856 | |||
AK166099.1 | - | 8 | 854 | |||
AK172038.1 | - | 8 | 854 | |||
AK172350.1 | - | 8 | 859 | |||
AK173141.1 | - | 8 | 806 | |||
AK181017.1 | - | 3 | 112 | |||
BC023301.1 | - | 8 | 795 | 3671787 | ||
BC058373.1 | - | 8 | 842 | 6828037 | ||
BC060036.1 | - | 8 | 790 | 6530155 |