Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aif1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145144.1 | NP_660126.1 | 4, 5 | 253 | ||
GenBank mRNA | AB035322.1 | - | 4, 5 | 205 | ||
AK028955.1 | - | 4, 5 | 211 | |||
AK045539.1 | - | 4, 5 | 283 | |||
BC024599.1 | - | 4, 5 | 253 | 5044495 |