Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4gat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175383.2 | NP_780592.1 | 2 | 1329 | ||
GenBank mRNA | AK050347.1 | - | 2 | 1303 | ||
AK075733.1 | - | 2 | 1210 | |||
AK083857.1 | - | 1 | 1526 | |||
AK158899.1 | - | 2 | 1330 | |||
BC002191.1 | - | 2 | 657 | |||
BC069927.1 | - | 2 | 1257 | 4010980 |
Gene Symbol: Brms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC038380.1 | - | 1264 |