Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110162.1 | NP_001103632.1 | 13 | 1902 | ||
NM_175384.4 | NP_780593.3 | 13 | 1605 | |||
GenBank mRNA | AK031313.1 | - | 9 | 1047 | ||
AK160223.1 | - | 13 | 1665 | |||
AK166952.1 | - | 13 | 1672 | |||
BC037136.1 | - | 13 | 1702 | |||
BC067009.1 | - | 13 | 1633 | |||
BC075682.1 | - | 13 | 1605 | |||
BC116298.1 | - | 9 | 1047 | 40095010 | ||
BC116299.1 | - | 9 | 1047 | 40095011 |