Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110162.1 | NP_001103632.1 | 6 | 1019 | ||
NM_175384.4 | NP_780593.3 | 6 | 722 | |||
GenBank mRNA | AK031083.1 | - | 6 | 785 | ||
AK031313.1 | - | 2 | 164 | |||
AK160223.1 | - | 6 | 781 | |||
AK166952.1 | - | 6 | 788 | |||
BC037136.1 | - | 6 | 819 | |||
BC067009.1 | - | 6 | 750 | |||
BC075682.1 | - | 6 | 722 | |||
BC116298.1 | - | 2 | 164 | 40095010 | ||
BC116299.1 | - | 2 | 164 | 40095011 |