Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d10c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178650.3 | NP_848765.2 | 2 | 105 | ||
GenBank mRNA | AK040487.1 | - | 2 | 106 | ||
AK075803.1 | - | 2 | 92 | |||
AK088119.1 | - | 170 | ||||
AK131177.1 | - | 1 | 86 | |||
AK146426.1 | - | 2 | 99 | |||
AK153836.1 | - | 2 | 103 | |||
AK154444.1 | - | 1 | 83 | |||
BQ829173.1 | - | 2 | 341 |