Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d10c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178650.3 | NP_848765.2 | 3 | 268 | ||
GenBank mRNA | AK040487.1 | - | 3 | 269 | ||
AK075803.1 | - | 3 | 255 | |||
AK088119.1 | - | 333 | ||||
AK131177.1 | - | 2 | 249 | |||
AK146426.1 | - | 3 | 262 | |||
AK153836.1 | - | 3 | 266 | |||
AK154444.1 | - | 2 | 246 | |||
BC094901.1 | - | 2 | 243 | |||
BQ829173.1 | - | 3 | 504 |