Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ciapin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134141.4 | NP_598902.1 | 4 | 494 | ||
GenBank mRNA | AK076116.1 | - | 4 | 450 | ||
AK079615.1 | - | 4 | 456 | |||
AK088757.1 | - | 3 | 421 | |||
AK136821.1 | - | 4 | 488 | |||
AK146282.1 | - | 4 | 442 | |||
AK163705.1 | - | 4 | 473 | |||
AK168252.1 | - | 4 | 444 | |||
AK178553.1 | - | 2 | 115 | |||
AK183681.1 | - | 2 | 115 | |||
AY523555.1 | - | 3 | 415 | |||
BC016261.1 | - | 3 | 412 | 4920560 | ||
BC021864.1 | - | 4 | 434 | 5123958 | ||
BC021949.1 | - | 3 | 245 |