Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ciapin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134141.4 | NP_598902.1 | 3, 4 | 425 | ||
GenBank mRNA | AK076116.1 | - | 3, 4 | 381 | ||
AK079615.1 | - | 3, 4 | 387 | |||
AK088757.1 | - | 2, 3 | 352 | |||
AK136821.1 | - | 3, 4 | 419 | |||
AK146282.1 | - | 3, 4 | 373 | |||
AK163705.1 | - | 3, 4 | 404 | |||
AK168252.1 | - | 3, 4 | 375 | |||
AK178553.1 | - | 1, 2 | 46 | |||
AK183681.1 | - | 1, 2 | 46 | |||
AK201030.1 | - | 1, 2 | 46 | |||
AY523555.1 | - | 2, 3 | 346 | |||
BC016261.1 | - | 2, 3 | 343 | 4920560 | ||
BC021864.1 | - | 3, 4 | 365 | 5123958 | ||
BC021949.1 | - | 2, 3 | 176 |