Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACYP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107.4 | NP_001098.1 | 3 | 318 | ||
NM_001302616.1 | NP_001289545.1 | 3 | 300 | |||
NM_001302617.1 | NP_001289546.1 | 3 | 496 | |||
NR_126393.1 | 4 | 397 | ||||
NR_126394.1 | 2 | 404 | ||||
NR_126395.1 | 2 | 226 | ||||
GenBank mRNA | AK312101.1 | - | 3 | 318 | ||
BC035568.1 | - | 3 | 303 | 5392248 | ||
BG614847.1 | - | 4 | 364 | |||
BP316920.1 | - | 3 | 281 | |||
BX104912.1 | - | 2 | 171 | |||
CD675334.1 | - | 3 | 505 | |||
DB071115.1 | - | 3 | 318 | |||
HY012032.1 | - | 2 | 366 | |||
X84194.1 | - | 2 | 282 |