Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exosc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175399.4 | NP_780608.1 | 3 | 1689 | ||
GenBank mRNA | AK005101.2 | - | 3 | 1617 | ||
AK083308.1 | - | 3 | 1650 | |||
AK089890.1 | - | 3 | 1690 | |||
AK144062.1 | - | 3 | 1604 | |||
AK167629.1 | - | 3 | 1652 |