Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc30a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178651.3 | NP_848766.2 | 14 | 1292 | ||
GenBank mRNA | AF263460.1 | - | 12 | 846 | ||
AK028885.1 | - | 14 | 1259 | |||
AK033990.1 | - | 317 | ||||
AK035541.1 | - | 14 | 1279 | |||
AK038559.1 | - | 13 | 1198 | |||
AK082853.1 | - | 14 | 1294 | |||
AK143277.1 | - | 14 | 1293 | |||
AK149549.1 | - | 6 | 3743 | |||
AK179008.1 | - | 6 | 380 | |||
AK182342.1 | - | 6 | 370 | |||
AK204464.1 | - | 5 | 311 | |||
AK205884.1 | - | 6 | 338 | |||
AY682914.1 | - | 15 | 1285 | |||
BC023833.1 | - | 6 | 396 | |||
BC024849.1 | - | 1 | 25 | |||
BC026565.1 | - | 10 | 710 | |||
BC027806.1 | - | 14 | 1054 | |||
BC031705.1 | - | 11 | 806 | |||
BC055773.1 | - | 11 | 745 | |||
BC078440.1 | - | 12 | 869 | |||
BC141008.1 | - | 15 | 1258 | 9055620 |