Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uhrf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144873.2 | NP_659122.2 | 4 | 949 | ||
GenBank mRNA | AB116653.1 | - | 4 | 684 | ||
AF274047.1 | - | 4 | 859 | |||
AK080925.1 | - | 4 | 990 | |||
AK082518.1 | - | 3 | 760 | |||
BC060241.1 | - | 4 | 949 | 6334856 |