Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Supt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178652.2 | NP_848767.1 | 7 | 797 | ||
GenBank mRNA | AK050321.1 | - | 7 | 644 | ||
AK075916.1 | - | 7 | 797 | |||
AK087935.1 | - | 7 | 642 | |||
BC051540.1 | - | 6 | 527 | |||
BC096051.1 | - | 7 | 581 |