Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sobp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175407.3 | NP_780616.4 | 6 | 1366 | ||
GenBank mRNA | BC057055.1 | - | 5 | 949 | ||
BC058972.1 | - | 1 | 228 | |||
BC059851.1 | - | 6 | 1366 | 5704623 | ||
DQ157775.1 | - | 6 | 925 |