Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sccpdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178653.3 | NP_848768.1 | 10 | 1199 | ||
GenBank mRNA | AK042919.1 | - | 10 | 1087 | ||
AK046033.1 | - | 10 | 1200 | |||
AK046468.1 | - | 10 | 1187 | |||
AK050554.1 | - | 10 | 1148 | |||
AK053698.1 | - | 1 | 444 | |||
AK077695.1 | - | 10 | 1149 | |||
AK082803.1 | - | 10 | 1084 | |||
AK086448.1 | - | 10 | 1222 | |||
AK086632.1 | - | 10 | 1198 | |||
AK166018.1 | - | 10 | 1111 | |||
AK212321.1 | - | 4 | 349 | |||
BC025803.1 | - | 10 | 1196 | 5100552 |