Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sccpdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178653.3 | NP_848768.1 | 3 | 482 | ||
GenBank mRNA | AK042919.1 | - | 3 | 370 | ||
AK046033.1 | - | 3 | 483 | |||
AK046468.1 | - | 3 | 469 | |||
AK050554.1 | - | 3 | 431 | |||
AK077695.1 | - | 3 | 432 | |||
AK081021.1 | - | 2 | 2491 | |||
AK082803.1 | - | 3 | 367 | |||
AK086448.1 | - | 3 | 505 | |||
AK086632.1 | - | 3 | 480 | |||
AK145399.1 | - | 3 | 466 | |||
AK166018.1 | - | 3 | 394 | |||
AK182882.1 | - | 3 | 224 | |||
AK215316.1 | - | 3 | 224 | |||
BC025803.1 | - | 3 | 479 | 5100552 |