Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290656.1 | NP_001277585.1 | 4 | 1039 | ||
NM_029924.2 | NP_084200.1 | 11 | 2103 | |||
GenBank mRNA | AK155574.1 | - | 11 | 2106 | ||
AK173176.1 | - | 4 | 970 | |||
BC050251.1 | - | 1 | 64 | |||
BC138375.1 | - | 4 | 944 | 8861395 | ||
BC138378.1 | - | 4 | 944 | 8861398 |