Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027321.3 | NP_081597.1 | 4 | 565 | ||
NM_175413.3 | NP_780622.1 | 4 | 565 | |||
GenBank mRNA | AK008119.1 | - | 4 | 533 | ||
AK035072.1 | - | 4 | 567 | |||
AK039277.1 | - | 4 | 562 | |||
AK046671.1 | - | 4 | 461 | |||
AK052159.1 | - | 4 | 517 | |||
AK079125.1 | - | 4 | 543 | |||
BC119529.2 | - | 4 | 514 | 40056120 | ||
BC119530.1 | - | 4 | 515 | 40056121 |