Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175414.4 | NP_780623.1 | 5 | 480 | ||
GenBank mRNA | AK079139.1 | - | 5 | 453 | ||
AK158729.1 | - | 4 | 365 | |||
AK163395.1 | - | 5 | 536 | |||
AK191038.1 | - | 2 | 143 | |||
AK199601.1 | - | 3 | 289 | |||
BC052503.1 | - | 5 | 462 | 5146854 |