Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mybpc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252372.1 | NP_001239301.1 | 5 | 651 | ||
NM_175418.5 | NP_780627.2 | 6 | 693 | |||
GenBank mRNA | AK132188.1 | - | 5 | 525 | ||
BC062158.1 | - | 6 | 555 | 30134842 | ||
HQ848554.1 | - | 8 | 541 |