Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: R3hdm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177994.4 | NP_818775.2 | 6 | 606 | ||
GenBank mRNA | AK081288.1 | - | 6 | 595 | ||
AK088107.1 | - | 6 | 607 | |||
AK139047.1 | - | 6 | 597 | |||
AK159869.1 | - | 6 | 584 | |||
AK165610.1 | - | 6 | 577 | |||
AK188300.1 | - | 3 | 200 | |||
BC034653.1 | - | 5 | 409 | |||
BC040686.1 | - | 3 | 238 | |||
BC095970.1 | - | 6 | 587 | 6530790 | ||
BC132463.1 | - | 6 | 682 | 40130740 | ||
BC138461.1 | - | 6 | 682 | 8861481 |