Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1qtnf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135172.1 | NP_001128644.1 | 2, 3 | 667 | ||
NM_175425.4 | NP_780634.2 | 2, 3 | 310 | |||
GenBank mRNA | AK017441.1 | - | 2, 3 | 679 | ||
AK078818.1 | - | 2, 3 | 473 | |||
BC038308.1 | - | 2, 3 | 310 | |||
BC090967.1 | - | 2, 3 | 648 | 30652983 | ||
DQ002400.1 | - | 2, 3 | 319 |