Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdcp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133974.3 | NP_598735.2 | 4 | 972 | ||
GenBank mRNA | AK087108.1 | - | 4 | 870 | ||
BC018410.1 | - | 4 | 983 | |||
BC034137.1 | - | 2 | 380 | |||
BC059937.1 | - | 4 | 879 | |||
BC085253.1 | - | 4 | 972 | 5347122 |