Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdcp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133974.3 | NP_598735.2 | 3 | 771 | ||
GenBank mRNA | AK035674.1 | - | 3 | 733 | ||
AK087108.1 | - | 3 | 669 | |||
BC018410.1 | - | 3 | 782 | |||
BC034137.1 | - | 1 | 179 | |||
BC059937.1 | - | 3 | 678 | |||
BC085253.1 | - | 3 | 771 | 5347122 |