Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam175b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198017.3 | NP_932134.3 | 9 | 978 | ||
GenBank mRNA | AK146064.1 | - | 9 | 953 | ||
AK165918.1 | - | 9 | 917 | |||
AK170704.1 | - | 9 | 953 | |||
AK171524.1 | - | 9 | 904 | |||
AK172903.1 | - | 2 | 1670 | |||
BC030838.1 | - | 6 | 734 |