Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ampd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289719.1 | NP_001276648.1 | 9, 10 | 1222 | ||
NM_001289720.1 | NP_001276649.1 | 8, 9 | 1039 | |||
NM_028779.5 | NP_083055.1 | 8, 9 | 1308 | |||
GenBank mRNA | AK004759.2 | - | 9, 10 | 1207 | ||
AK034844.1 | - | 8, 9 | 1012 | |||
AK169980.1 | - | 9, 10 | 1208 | |||
BC049119.1 | - | 8, 9 | 1269 | 6812571 |