Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ank2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034168.1 | NP_001029340.1 | 4 | 2029 | ||
NM_178655.3 | NP_848770.2 | 19 | 2332 | |||
GenBank mRNA | AK032060.1 | - | 19 | 2332 | ||
AK083826.1 | - | 4 | 588 | |||
BC043123.1 | - | 4 | 2029 | |||
BC059251.1 | - | 19 | 2283 | 6405358 |