Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hyal3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178020.3 | NP_821139.2 | 2 | 599 | ||
GenBank mRNA | AF074489.1 | - | 1 | 555 | ||
AY048681.1 | - | 2 | 588 | |||
BC018457.1 | - | 2 | 599 | 3592874 |