Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134156.2 | NP_598917.1 | 20, 21 | 2800 | ||
GenBank mRNA | AK014073.1 | - | 1, 2 | 108 | ||
AK142591.1 | - | 11, 12 | 1517 | |||
AK154281.1 | - | 20, 21 | 2793 | |||
BC003232.1 | - | 8, 9 | 1045 | |||
BC023206.1 | - | 7, 8 | 1002 | |||
BC038919.1 | - | 3, 4 | 655 | |||
BC054830.1 | - | 20, 21 | 2745 | 6308412 | ||
DQ288940.1 | - | 20, 21 | 2505 |