Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyb5r3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029787.2 | NP_084063.1 | 9 | 833 | ||
GenBank mRNA | AF332059.1 | - | 9 | 757 | ||
AF332060.1 | - | 9 | 757 | |||
AK002640.1 | - | 9 | 805 | |||
AK128917.1 | - | 9 | 827 | |||
AK213922.1 | - | 5 | 331 | |||
BC004760.1 | - | 9 | 840 | 3154531 | ||
BC032013.1 | - | 9 | 815 | 4238275 | ||
BC043074.1 | - | 9 | 833 | 6404746 |