Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163746.1 | NP_001157218.1 | 6 | 730 | ||
NM_028352.4 | NP_082628.3 | 6 | 730 | |||
GenBank mRNA | AK013402.1 | - | 6 | 713 | ||
AK028066.1 | - | 6 | 720 | |||
AK032776.1 | - | 6 | 682 | |||
AK036561.1 | - | 6 | 716 | |||
AK043547.1 | - | 6 | 731 | |||
AK049337.1 | - | 6 | 682 | |||
AK049758.1 | - | 6 | 717 | |||
AK051706.1 | - | 6 | 670 | |||
AK153501.1 | - | 5 | 550 | |||
AK160913.1 | - | 6 | 731 | |||
AK165513.1 | - | 6 | 715 | |||
AK169082.1 | - | 6 | 695 | |||
AK169141.1 | - | 6 | 689 | |||
AK169787.1 | - | 6 | 704 | |||
BC138700.1 | - | 6 | 655 | 8861722 |